Learn More
CAP-D2 Rabbit anti-Human, Polyclonal, Bethyl Laboratories
Rabbit Polyclonal Antibody
Supplier: Bethyl Laboratories, Inc A300601A

Description
The recommended shelf life for this product is 1 year from date of receipt.
NCAPD2 is the regulatory subunit of the condensin complex, a complex required for conversion of interphase chromatin into mitotic-like condense chromosomes. The condensin complex probably introduces positive supercoils into relaxed DNA in the presence of type I topoisomerases and converts nicked DNA into positive knotted forms in the presence of type II topoisomerases. NCAPD2 may target the condensin complex to DNA via its C-terminal domain.Specifications
CAP-D2 | |
Polyclonal | |
Unconjugated | |
NCAPD2 | |
Non-SMC condensin I complex, subunit D2, CAP-D2, CNAP1, KIAA0159, hCAP-D2 | |
Rabbit | |
Antigen affinity chromatography | |
RUO | |
9918 | |
4° C | |
Liquid |
Immunoprecipitation, Western Blot | |
0.2 mg/ml | |
TBS with 0.1% BSA and 0.09% sodium azide | |
Q15021 | |
NCAPD2 | |
Between 1350 and C-term | |
100 μL | |
Primary | |
Human | |
Antibody | |
IgG |
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.