Learn More
CAP-G Rabbit anti-Human, Polyclonal, Bethyl Laboratories
Rabbit Polyclonal Antibody
Supplier: Bethyl Laboratories, Inc A300602A
Description
The recommended shelf life for this product is 1 year from date of receipt.
This gene encodes a subunit of the condensin complex, which is responsible for the condensation and stabilization of chromosomes during mitosis and meiosis. Phosphorylation of the encoded protein activates the condensin complex. There are pseudogenes for this gene on chromosomes 8 and 15. Alternative splicing results in multiple transcript variants.Specifications
| CAP-G | |
| Polyclonal | |
| Unconjugated | |
| NCAPG | |
| CAPG, CHCG, FLJ12450, HCAP-G, MGC126525, NY-MEL-3 | |
| Rabbit | |
| Antigen affinity chromatography | |
| RUO | |
| 64151 | |
| 4° C | |
| Liquid |
| Immunoprecipitation, Western Blot | |
| 0.2 mg/ml | |
| TBS with 0.1% BSA and 0.09% sodium azide | |
| Q9BPX3 | |
| NCAPG | |
| Between 950 and C-term | |
| 100 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.