Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Simport Cap with O-Ring and Long Lip Seal

Catalog No. 23045749 Shop All Simport Scientific Products
Change view
Click to view available options
Color:
Clear
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Color
23-045-749 Clear
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 23-045-749 Supplier Simport Supplier No. T505N
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Cap made of polypropylene with a long skirt and super fast thread design for easy seal.

  • Caps are available either with a silicone washer between the cap and the tube to ensure a positive leakproof seal at all temperatures.
  • As for tubes, these closures are also made of polypropylene, providing the same coefficient of expansion for both, which further enhances the leakproof qualities of the vials at changing temperatures.
  • The cap features a long skirt and a super-fast thread design that allows it to be removed or sealed with a single turn.

Specifications

Liner Material Linerless
Pre-sterilized No
Product Type Screw Cap
Includes O-ring, Long Lip Seal
Autoclavable Autoclavable
Material Polypropylene
Color Clear
Sterility Non-sterile
For Use With (Equipment) Tubes
Fit for Purpose Certified Quality
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.