Learn More
Description
- Non-Pyrogenic
- Domed lid
- Highly transparent polypropylene
- Free of detectable DNase, RNase, human DNA
- Thin wall for optimal heat transfer
- Low evaporation rate in PCR
- Seals 8-tube strips in addition to
Specifications
Specifications
| Color | Violet |
| Quantity | 1250/Cs. |
| For Use With (Equipment) | PCR 8-tube Strip |
| Sterility | Non-sterile |
| Material | Polypropylene |
| Product Type | Cap Strip |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.