Learn More
Invitrogen™ CAP1 Polyclonal Antibody

Description
Immunogen sequence: MADMQNLVER LERAVGRLEA VSHTSDMHRG YADSPSKAGA APYVQAFDSL LAGPVAEYLK ISKEIGGDVQ KHAEMVHTGL KLERALLVTA SQCQQPAENK; Positive Samples: HeLa, 293T, A-431, HepG2, Rat thymus, Mouse thymus; Cellular Location: Cell membrane, Peripheral membrane protein.
Specifications
Specifications
| Antigen | CAP1 |
| Applications | ELISA, Western Blot |
| Classification | Polyclonal |
| Concentration | 0.44 mg/mL |
| Conjugate | Unconjugated |
| Formulation | PBS with 50% glycerol and 0.01% thimerosal; pH 7.3 |
| Gene | CAP1 |
| Gene Accession No. | P40124, Q01518, Q08163 |
| Gene Alias | adenylate cyclase associated protein 1; adenylyl cyclase-associated CAP protein, yeast homolog 1; adenylyl cyclase-associated protein 1; CAP; CAP 1; CAP, adenylate cyclase-associated protein 1; CAP, adenylate cyclase-associated protein 1 (yeast); CAP1; CAP1-PEN; cyclase-associated protein homologue; Mch1; RP11-115D7.1; testis secretory sperm-binding protein Li 218p |
| Gene Symbols | CAP1 |
| Show More |
Safety and Handling
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.