Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ CAP2 Polyclonal Antibody
Rabbit Polyclonal Antibody
Supplier: Invitrogen™ PA597533
Description
CAP2 Polyclonal Antibody for Western Blot, IHC (P), ELISA
This gene was identified by its similarity to the gene for human adenylyl cyclase-associated protein. The function of the protein encoded by this gene is unknown. However, the protein appears to be able to interact with adenylyl cyclase-associated protein and actin.
Specifications
CAP2 | |
Polyclonal | |
Unconjugated | |
Cap2 | |
2810452G09Rik; Adenylyl cyclase-associated protein 2; AV045235; CAP 2; CAP, adenylate cyclase-associated protein, 2 (yeast); CAP2 | |
Rabbit | |
Protein G | |
RUO | |
10486, 67252 | |
-20°C or -80°C if preferred | |
Liquid |
ELISA, Immunohistochemistry (Paraffin), Western Blot | |
2 mg/mL | |
PBS with 50% glycerol and 0.03% ProClin 300; pH 7.4 | |
P40123, Q9CYT6 | |
Cap2 | |
Recombinant Human Adenylyl cyclase-associated protein 2 protein (221-477AA). | |
100 μg | |
Primary | |
Human, Mouse | |
Antibody | |
IgG |
Safety and Handling
WARNING: Cancer - www.P65Warnings.ca.gov
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction