Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
CAPC/ LRRC26 Antibody, Novus Biologicals™

Rabbit Polyclonal Antibody
Supplier: Novus Biologicals NBP23087725UL
Description
CAPC / LRRC26 Polyclonal specifically detects CAPC / LRRC26 in Human samples. It is validated for Immunohistochemistry, Immunohistochemistry-Paraffin.Specifications
CAPC / LRRC26 | |
Polyclonal | |
Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunohistochemistry-Paraffin 1:50 - 1:200 | |
Q2I0M4 | |
LRRC26 | |
This antibody was developed against a recombinant protein corresponding to amino acids: PEVCTCVPGGLASCSALSLPAVPPGLSLRLRALLLDHNRVRA | |
25 μL | |
Primary | |
Specificity of human CAPC LRRC26 antibody verified on a Protein Array containing target protein plus 383 other non-specific proteins. | |
Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles. |
Immunohistochemistry, Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin) | |
Unconjugated | |
PBS (pH 7.2) and 40% Glycerol with 0.02% Sodium Azide | |
CAPC, cytokeratin associated protein, leucine rich repeat containing 26 | |
Rabbit | |
Affinity Purified | |
RUO | |
389816 | |
Human | |
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction