Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
CapG Antibody (1D10), Alexa Fluor™ 488, Novus Biologicals™

Mouse Monoclonal Antibody
Supplier: Novus Biologicals NBP259450AF488
This item is not returnable.
View return policy
Description
CapG Monoclonal antibody specifically detects CapG in Human samples. It is validated for Western Blot, ELISA, ImmunofluorescenceSpecifications
CapG | |
Monoclonal | |
Alexa Fluor 488 | |
actin-regulatory protein CAP-G, AFCP, capping protein (actin filament), gelsolin-like, gelsolin-like capping protein, macrophage capping protein, macrophage-capping protein, MCPActin regulatory protein CAP-G | |
Recombinant human CAPG (1-348aa) purified from E. coli | |
0.1 mL | |
Cell Biology, Cellular Markers | |
822 | |
Store at 4°C in the dark. | |
IgG2b κ |
Western Blot, ELISA, Immunofluorescence | |
1D10 | |
50mM Sodium Borate | |
Mouse | |
Protein A purified | |
RUO | |
Primary | |
Human | |
Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction