Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Capicua Antibody, Alexa Fluor™ 350, Novus Biologicals™

Rabbit Polyclonal Antibody
Supplier: Novus Biologicals NB11059905AF350
Description
Capicua Polyclonal specifically detects Capicua in Human, Mouse, Rat, Canine samples. It is validated for Immunocytochemistry/Immunofluorescence.Specifications
Capicua | |
Polyclonal | |
Immunocytochemistry/Immunofluorescence | |
capicua homolog (Drosophila), KIAA0306capicua (Drosophila) homolog, protein capicua homolog | |
Rabbit | |
0.1 mL | |
23152 | |
Store at 4°C in the dark. |
Immunocytochemistry, Immunofluorescence | |
Alexa Fluor 350 | |
50 mM sodium borate with 0.05% sodium azide | |
CIC | |
The epitope recognized by this antibody maps to a region within residue 1150-1200 of human Capicua. [NP_055940.3] | |
Primary | |
Human, Mouse, Rat, Canine | |
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction