Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Capicua Antibody, Alexa Fluor™ 700, Novus Biologicals™

Rabbit Polyclonal Antibody
Supplier: Novus Biologicals NB11059906AF700
Description
Capicua Polyclonal specifically detects Capicua in Human, Mouse, Rat, Canine samples. It is validated for Western Blot, Immunocytochemistry/Immunofluorescence, Knockout Validated.Specifications
Capicua | |
Polyclonal | |
Western Blot, Immunocytochemistry/Immunofluorescence, Knockout Validated | |
Q96RK0 | |
CIC | |
The epitope recognized by this antibody maps to a C-terminal region within residue 1500-1608 of human Capicua. [NP_055940.3] | |
Affinity Purified | |
RUO | |
Primary | |
Human, Mouse, Rat, Canine | |
IgG |
Western Blot, Immunocytochemistry, Immunofluorescence, KnockDown | |
Alexa Fluor 700 | |
50mM Sodium Borate with 0.05% Sodium Azide | |
capicua homolog (Drosophila), KIAA0306capicua (Drosophila) homolog, protein capicua homolog | |
Rabbit | |
250 kDa | |
0.1 mL | |
Cancer, Neuroscience | |
23152 | |
Store at 4C in the dark. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction