Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Capicua Antibody, DyLight 405, Novus Biologicals™

Rabbit Polyclonal Antibody
Supplier: Novus Biologicals NB11059905V
Description
Capicua Polyclonal specifically detects Capicua in Human, Mouse, Rat, Canine samples. It is validated for Immunocytochemistry/Immunofluorescence, Western Blot (Negative).Specifications
Capicua | |
Polyclonal | |
DyLight 405 | |
50mM Sodium Borate with 0.05% Sodium Azide | |
capicua homolog (Drosophila), KIAA0306capicua (Drosophila) homolog, protein capicua homolog | |
Rabbit | |
164 kDa | |
0.1 mL | |
Cancer, Neuroscience | |
23152 | |
Store at 4C in the dark. |
Immunocytochemistry, Immunofluorescence, Western Blot | |
1mg/mL | |
Immunocytochemistry/Immunofluorescence, Western Blot (Negative) | |
Q96RK0 | |
CIC | |
The epitope recognized by this antibody maps to a region within residue 1150-1200 of human Capicua. [NP_055940.3] | |
Affinity Purified | |
RUO | |
Primary | |
Human, Mouse, Rat, Canine | |
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction