Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Capicua Antibody, mFluor Violet 610 SE, Novus Biologicals™

Rabbit Polyclonal Antibody
Supplier: Novus Biologicals NB11059906MFV610
Description
Capicua Polyclonal antibody specifically detects Capicua in Human, Mouse, Rat, Canine samples. It is validated for Western Blot, Immunocytochemistry/ Immunofluorescence, Immunoprecipitation, KnockDownSpecifications
Capicua | |
Polyclonal | |
Western Blot, Immunocytochemistry/ Immunofluorescence, Immunoprecipitation, Knockout Validated | |
capicua homolog (Drosophila), KIAA0306capicua (Drosophila) homolog, protein capicua homolog | |
The epitope recognized by this antibody maps to a C-terminal region within residue 1500-1608 of human Capicua. [NP_055940.3] | |
0.1 mL | |
Cancer, Neuroscience | |
23152 | |
Store at 4C in the dark. | |
IgG |
Western Blot, Immunocytochemistry, Immunoprecipitation, KnockDown | |
mFluor Violet 610 SE | |
50mM Sodium Borate | |
Rabbit | |
Immunogen affinity purified | |
RUO | |
Primary | |
Human, Mouse, Rat, Canine | |
Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction