Learn More
MilliporeSigma™ Supelco™ Capillary Cleaving™ Tools and Accessories
Increases column performance by making the properly sized, squarely-cut ferrule to seal capillary column ends
$160.60
Specifications
For Use With (Application) Making Scalpel-Like Cuts to Polyimide-Coated Fused Silica
Material Sapphire Cutting Blade
View More Specs
Products 1
Catalog Number Mfr. No. Type Price Quantity & Availability  
Catalog Number Mfr. No. Type Price Quantity & Availability  
11-102-0881
View Documents
MilliporeSigma™ Supelco™
23740U
Fixed Blade Version
Each of 1 for $160.60
Only null left
Add to Cart
Description
Description
  • Makes scalpel-like cuts to polyimide-coated fused silica
  • Leaves no jagged edges to create problems
  • Industrial sapphire cutting edges remains sharp indefinitely
  • Spring-loaded retractable blade reduces the chance of breakage if the tool is dropped
  • Replacement sapphire cutting blades are available
Specifications
Specifications
Making Scalpel-Like Cuts to Polyimide-Coated Fused Silica
Sapphire Cutting Blade
Videos
SDS
Documents
Documents
Product Certifications
Cleaving is a trademark of Sigma-Aldrich Co. LLC
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.