Learn More
Restek™ Capillary Dedicated FID Replacement Jet
SDP
Catalog No. 06611260 Shop All Restek Corporation Products
Encompass_Preferred
Change view
Click to view available options
For Use With (Equipment):
Agilent 5890/6850/6890/7890 GCs
Catalog No. For Use With (Equipment)
06-611-260 Agilent 5890/6850/6890/7890 GCs
1 options
Catalog No. 06-611-260 Supplier Restek™ Supplier No. 23078
High Temperature Version, special processing ensures the highest degree of cleanliness
  • Hight Temperature Version
  • Threads specially coated for easy installation and removal
  • Special processing ensures the highest degree of cleanliness
  • Meets or exceeds original manufacturer's performance
  • Siltek Treated
  • Extremely inert
  • For use with active compounds
Specifications
Type High Temperature
Length (Metric) 43 mm
Material Siltek treated
Diameter (English) Inner 0.018 in.
Quantity 1/Pk.
For Use With (Equipment) Agilent 5890/6850/6890/7890 GCs