Learn More
3M™ DBI-Sala™ Delta™ Vest Style Harnesses
Catalog No. 19066800 Shop All 3M Products
Change view
Click to view available options
Size:
Universal
X-Large
D-Ring Location:
1 back
1 back, 2 hip
Back, Side
Buckle Style (Leg):
Pass through
Pass-Through
Tongue Buckle
Style:
Construction Style, w/ loops for belt
Vest Style
5 product options available for selection
Product selection table with 5 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Size D-Ring Location Buckle Style (Leg) Style
19-066-800 Universal 1 back, 2 hip Tongue Buckle Vest Style
19-066-803 Universal 1 back, 2 hip Pass-Through Vest Style
19-066-884 X-Large Back, Side Tongue Buckle Vest Style
19-066-804 Universal 1 back Pass through Vest Style
18-706 Universal 1 back, 2 hip Pass-Through Construction Style, w/ loops for belt
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 5 options available.
5 options
Catalog No. 19-066-800 Supplier 3M™ Supplier No. 7012691269
Add to Cart
Add to Cart
No-Tangle™ triangular design creates frame that allows the harness to fall into place when donned.
  • Spring-loaded stand-up dorsal D-ring design ensures fast, easy and safe connections
  • Rip-stitch indicators allow to inspect the harness at glance, for prior damaging impact loads
  • Harness falls into place, allowing quick and easy donning
  • Side D-rings for positioning
  • Adjustable non-slip chest strap with easy to use pass-thru buckle
  • Tongue buckle leg straps for ease of use and will not loose
  • Lanyard keepers holds unused snap hooks and help reduce trip hazard
  • Revolver vertical torso adjuster provides simple and fast adjustment eliminating loose ends and locks webbing into place preventing slippage
  • Velcro cover protects harness labels and holds I-Safe™ ID tag
Compliance
Meets ANSI™ Z359.11 requirements.
TRUSTED_SUSTAINABILITY
Specifications
Material Polyester Webbing
Style Vest Style
D-Ring Location 1 back, 2 hip
Buckle Style (Chest) Pass-Through
Buckle Style (Leg) Tongue Buckle
Size Universal