Learn More
3M™ DBI-Sala™ ExoFit NEX™ Vest-Style Positioning/Climbing Harness
Catalog No. 19810558 Shop All 3M Products
Change view
Click to view available options
Size:
Small
Medium
Large
X-Large
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Size
19-810-558 Medium
19-810-557 Small
19-810-559 Large
19-810-560 X-Large
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
Catalog No. 19-810-558 Supplier 3M™ Supplier No. 7012776902/DEL
For comfort, security and no-tangle donning.
  • Hybrid shoulder, back and leg comfort padding
  • Stand-up Tech-Lite aluminum back D-ring
  • Tech-Lite aluminum front D-ring
  • Tech-Lite aluminum side D-rings
  • Duo-Lok quick connect chest and leg straps
  • Repel webbing
  • Protective shoulder caps
  • Loops for body belt
  • Built-in lanyard keepers
  • Revolver vertical torso adjusters
  • Impact Indicator
  • Integrated trauma straps
  • Built-in reflective materials
  • Protected labels, equipped with i-Safe
Compliance
Meets OSHA 1910.66 and OSHA 1926.502; ANSI A10.32, ANSI Z359.1, ANSI Z359.3 and ANSI Z359.4 requirements.
TRUSTED_SUSTAINABILITY
Specifications
Size Medium