Description
ExoFit NEX Construction Style Harness with aluminum front, back and side D-rings, locking quick connect buckles, sewn-in hip pad and removable belt.
- Wraps around for the ultimate in no-tangle donning and comfortable security
- Shoulder, hip and leg padding is built-in so it can't slip
- Breathable lining guarantees stay-dry and comfortable every day
- Memory-fit features allowing user to lock the adjustments in place to prevent the web from slipping
Compliance
Meets OSHA 1910.66 and OSHA 1926.502; ANSI™ A10.32, ANSI Z359.1, ANSI Z359.3 and ANSI Z359.4 requirements.
Specifications
Specifications
| Material | Aluminum |
| Style | Construction Style |
| Size | Small |
| D-Ring Location | Back, Front, Side |