Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Thermo Scientific™ Capitol Vial Transport-Tube Screw Caps

Catalog No. 1437692
Change view
Click to view available options
Color:
Red
White
No. per Case:
2000/Cs.
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Color No. per Case
14-376-92 White 2000/Cs.
14-376-100 Red 2000/Cs.
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Catalog No. 14-376-92 Supplier Thermo Scientific™ Supplier No. S40201
This item is not returnable. View return policy

Thermo Scientific™ Capitol Vial Transport-Tube Screw Caps are made of polyethylene.

Thermo Scientific™ Capitol Vial Transport-Tube Screw Caps are made of polyethylene.

  • For 10 and 12 mL serum transport tubes
TRUSTED_SUSTAINABILITY

Specifications

Product Type Screw Cap
Diameter (Metric) Outer 15 mm
Material Polyethylene
Color White
Height (Metric) 96 mm
No. per Case 2000/Cs.
For Use With (Equipment) Capitol Vial 10 and 12 mL Transport Tubes (not included)
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.