Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ CAPN14 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA570519
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA570519 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA570519 Supplier Invitrogen™ Supplier No. PA570519
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

CAPN14 Polyclonal Antibody for Western Blot

Calpains are a family of cytosolic calcium-activated cysteine proteases involved in a variety of cellular processes including apoptosis, cell division, modulation of integrin-cytoskeletal interactions, and synaptic plasticity. CAPN14 belongs to the calpain large subunit family.
TRUSTED_SUSTAINABILITY

Specifications

Antigen CAPN14
Applications Western Blot
Classification Polyclonal
Concentration 0.5 mg/mL
Conjugate Unconjugated
Formulation PBS with 2% sucrose and 0.09% sodium azide
Gene CAPN14
Gene Accession No. A8MX76
Gene Alias Calcium-activated neutral proteinase 14; calpain 14; calpain-14; CANP 14; CAPN14
Gene Symbols CAPN14
Host Species Rabbit
Immunogen synthetic peptide directed towards the middle region of human CAPN14.
Purification Method Affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 440854
Target Species Human
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.