Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
CAPN9 Monoclonal Antibody (OTI3G10), TrueMAB™, OriGene

Mouse Monoclonal Antibody
Supplier: OriGene TA503664S


Description
CAPN9 Monoclonal antibody specifically detects CAPN9 in Human samples. It is validated for Immunohistochemistry (Paraffin), Western BlotSpecifications
CAPN9 | |
Monoclonal | |
0.8 mg/mL | |
PBS with 1% BSA, 50% glycerol and 0.02% sodium azide | |
Q9GZT8 | |
Mouse | |
Affinity chromatography | |
RUO | |
60491 | |
-20° C, Avoid Freeze/Thaw Cycles | |
IgG2b |
Immunohistochemistry (Paraffin), Western Blot | |
OTI3G10 | |
Unconjugated | |
NIF3L1 | |
2200003B16Rik; calpain 9; calpain-9; Capn9; Digestive tract-specific calpain; GC36; NCL4; nCL-4; New calpain 4; novel calpain large subunit-4; Protein CG36 | |
Full length human recombinant protein of human NIF3L1 produced in HEK293T cell. | |
30 μL | |
Primary | |
Human | |
Liquid |
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction