Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Honeywell Burdick & Jackson™ Capping Reagent, 84/16 THF/NMI (v/v)

Catalog No. p-7161168 Shop All Honeywell Research Chemicals Products
Click to view available options
Quantity:
4 x 2 L
4 x 4 L
Packaging Type:
Glass Bottle
This item is not returnable. View return policy

Catalog No. 60-048-384

Supplier: Honeywell Burdick & Jackson™ BR6502

Only null left
Add to Cart
Add to Cart
This item is not returnable. View return policy

BioSyn™, for DNA/RNA synthesis

Burdick & Jackson™ BioSyn™ reagents & solvents for DNA/RNA synthesis

  • Honeywell Burdick & Jackson™ BioSyn™ Solvents and Reagents are specifically processed and purified to exacting specifications, ensuring outstanding synthesis efficiency
  • ideal for oligonucleotide synthesis regardless of the application
  • All solvents and reagents are filtered (0.2-micron for oxidation reagents and 0.1-micron for all other reagents) to minimize particulate contamination
TRUSTED_SUSTAINABILITY

Specifications

CAS 616-47-7
Content And Storage Room Temperature
Density 0.910 g/cm3
Packaging Type Glass Bottle
Quantity 4 x 2 L
Form Liquid
WARNING: Cancer - www.P65Warnings.ca.gov
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.