Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Capping Reagent, Honeywell Burdick & Jackson™
Shop All Honeywell Research Chemicals Products
Click to view available options
Quantity:
20 L
56 L
200 L
Specifications
Specifications
| CAS | 75-05-8, 616-47-7 |
| CAS Max % | 75.00%, 25.00% |
| Boiling Point | 86.6°C.The physical data is that of the main component. |
| Quantity | 56 L |
| Flash Point | 6°C.The physical data is that of the main component. |
| Packaging | Returnable Stainless Steel Drum |
| Physical Form | Liquid, clear |
| Purity Grade Notes | Capping reagents |
Safety and Handling
WARNING: Cancer - www.P65Warnings.ca.gov
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction