Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Cappuccino Rabbit anti-Human, Polyclonal, Bioss
SDP

Catalog No. 501989432
Encompass_Preferred
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
50-198-9432 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 50-198-9432 Supplier Bioss Supplier No. BS9895R
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

The BLOC-1 complex is required for normal biogenesis of lysosome-related organelles, such as platelet dense granules and melanosomes. Plays a role in intracellular vesicle trafficking.

Specifications

Antigen Cappuccino
Applications Immunofluorescence, Immunohistochemistry (Paraffin), Western Blot
Classification Polyclonal
Concentration 1 μg/mL
Conjugate Unconjugated
Formulation PBS with 1% BSA, 50% glycerol and 0.09% sodium azide; pH
Gene BLOC1S4
Gene Accession No. Q9NUP1
Gene Alias BCAS4L; biogenesis of lysosomal organelles complex 1 subunit 4; biogenesis of lysosomal organelles complex-1, subunit 4, cappuccino; Biogenesis of lysosome-related organelles complex 1 subunit 4; BLOC-1 subunit 4; BLOC1S4; BLOS4; cappuccino homolog; CNO; Protein cappuccino homolog
Gene Symbols BLOC1S4
Host Species Rabbit
Immunogen KLH conjugated synthetic peptide derived from human CNO/Cappuccino.
Purification Method Protein A
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 55330
Target Species Human
Content And Storage -20°C
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.