Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Capreomycin sulfate, 90%
Click to view available options
Quantity:
250 mg
1 g
Description
Thought to inhibit protein synthesis by inhibiting normal ribosomal functions
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
- Thought to inhibit protein synthesis by inhibiting normal ribosomal functions
Specifications
Specifications
| Type | Capreomycin Sulfate |
| Physical Form | Powder |
| Color | White |
| Quantity | 1 g |
| Flash Point | >110°C (230°F) |
| Boiling Point | 1377°C |
| Solubility Information | Freely soluble in water |
| Formula Weight | 766.78 |
| Percent Purity | 90% |
| Chemical Name or Material | Capreomycin sulfate |
Safety and Handling
| Hazard Category | H302+H312+H332-H360 |
| Hazard Statement | GHS H Statement H360-H302-H312-H332 May damage fertility or the unborn child. Harmful if swallowed. Harmful in contact with skin. Harmful if inhaled. |
| Precautionary Statement | P201-P202-P261-P264b-P270-P271-P281-P301+P312-P302+P352-P304+P340-P308+P313-P312-P330-P363-P501c |
| EINECSNumber | 215-776-8 |
| RTECSNumber | EX8930000 |
| TSCA | No |
| storageNote1 | -30° to -10°C |
| Recommended Storage | Store at -20°C |
RUO – Research Use Only
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction