Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
CAPRIN1 Mouse anti-Human, Mouse, Rat, Clone: 6B10F11, Proteintech

Mouse Monoclonal Antibody
$518.60
Specifications
Antigen | CAPRIN1 |
---|---|
Clone | 6B10F11 |
Concentration | 0.74 mg/mL |
Applications | Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin), Western Blot |
Classification | Monoclonal |
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
50-173-6783
![]() ![]() |
Proteintech Group Inc
663521IG150UL |
150 μL |
Each of 1 for $518.60
|
|
|||||
Description
GPIP137 may regulate the transport and translation of mRNAs of proteins involved in synaptic plasticity in neurons and cell proliferation and migration in multiple cell types. It is expressed in the cytoplasm.Specifications
CAPRIN1 | |
0.74 mg/mL | |
Monoclonal | |
Liquid | |
RUO | |
PBS with 50% glycerol and 0.02% sodium azide; pH 7.3 | |
caprin 1, CAPRIN1, GPI anchored protein p137, GPI p137, GPIAP1, GPIP137, M11S1, p137GPI, RNG105 | |
Caprin1 | |
IgG1 | |
Protein G | |
Antibody |
6B10F11 | |
Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin), Western Blot | |
Unconjugated | |
Mouse | |
Human, Mouse, Rat | |
Q14444, Q5M9G3, Q60865 | |
362173, 4076, 53872 | |
CAPRIN1 Fusion Protein Ag7413 | |
Primary | |
-20°C | |
CAPRIN1 |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title