Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

CAPRIN1 Polyclonal Antibody, Bethyl Laboratories

Catalog No. 501566313
Encompass_Preferred
Change view
Click to view available options
Quantity:
20 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
50-156-6313 20 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 50-156-6313 Supplier Bethyl Laboratories, Inc Supplier No. A303882AT
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

The recommended shelf life for this product is 1 year from date of receipt. Based on 100% sequence identity, this antibody is predicted to react with Rat and Bovine

GPIP137 may regulate the transport and translation of mRNAs of proteins involved in synaptic plasticity in neurons and cell proliferation and migration in multiple cell types. It is expressed in the cytoplasm.

Specifications

Antigen CAPRIN1
Applications Western Blot
Classification Polyclonal
Conjugate Unconjugated
Formulation TBS with 0.1% BSA and 0.09% sodium azide
Gene CAPRIN1
Gene Accession No. Q14444, Q60865
Gene Alias Caprin-1; Cytoplasmic activation- and proliferation-associated protein 1; GPI-anchored membrane protein 1; GPI-anchored protein p137; p137GPI; Membrane component chromosome 11 surface marker 1; CAPRIN1; GPIAP1; GPIP137; M11S1
Gene Symbols CAPRIN1
Host Species Rabbit
Immunogen Between 550 and 600
Purification Method Antigen affinity chromatography
Quantity 20 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 4076, 53872
Target Species Human, Mouse
Content And Storage 4° C
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.