Learn More
CAPRIN1 Rabbit anti-Human, Mouse, Polyclonal, Bethyl Laboratories
Rabbit Polyclonal Antibody
Supplier: Bethyl Laboratories, Inc A303881A
Description
The recommended shelf life for this product is 1 year from date of receipt.
GPIP137 may regulate the transport and translation of mRNAs of proteins involved in synaptic plasticity in neurons and cell proliferation and migration in multiple cell types. It is expressed in the cytoplasm.Specifications
| CAPRIN1 | |
| Polyclonal | |
| Unconjugated | |
| CAPRIN1 | |
| Caprin-1; Cytoplasmic activation- and proliferation-associated protein 1; GPI-anchored membrane protein 1; GPI-anchored protein p137; p137GPI; Membrane component chromosome 11 surface marker 1; CAPRIN1; GPIAP1; GPIP137; M11S1 | |
| Rabbit | |
| Antigen affinity chromatography | |
| RUO | |
| 4076, 53872 | |
| 4° C | |
| Liquid |
| Immunoprecipitation, Western Blot | |
| 1 mg/ml | |
| phosphate, tris citrate with 0.09% sodium azide; pH 7-8 | |
| Q14444, Q60865 | |
| Caprin1 | |
| Between 450 and 500 | |
| 100 μL | |
| Primary | |
| Human, Mouse | |
| Antibody | |
| IgG |
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.