Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ CAPRIN2 Polyclonal Antibody
Rabbit Polyclonal Antibody
Supplier: Invitrogen™ PA5100421
Description
Antibody detects endogenous levels of total CAPRIN2.
The protein encoded by this gene may regulate the transport of mRNA. It may play a role in the differentiation of erythroblasts. Multiple transcript variants encoding different isoforms have been found for this gene.
Specifications
CAPRIN2 | |
Polyclonal | |
Unconjugated | |
Caprin2 | |
C1q domain containing 1; C1q domain-containing protein 1; C1QDC1; caprin family member 2; caprin2; caprin-2; Caprin-2-like protein; Cytoplasmic activation/proliferation-associated protein 2; EEG1; EEG-1; gastric cancer multidrug resistance-associated protein; KIAA1873; Protein EEG-1; RNA granule protein 140; RNG140 | |
Rabbit | |
Affinity Chromatography | |
RUO | |
232560, 65981, 686779 | |
-20°C | |
Liquid |
Western Blot | |
1 mg/mL | |
PBS with 50% glycerol and 0.02% sodium azide; pH 7.4 | |
Q05A80, Q6IMN6 | |
Caprin2 | |
A synthesized peptide derived from human CAPRIN2(Accession Q6IMN6), corresponding to amino acid residues F920-M970. | |
100 μL | |
Primary | |
Human, Mouse, Rat | |
Antibody | |
IgG |
Safety and Handling
WARNING: Cancer - www.P65Warnings.ca.gov
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction