Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ CAPRIN2 Polyclonal Antibody
Rabbit Polyclonal Antibody
Supplier: Invitrogen™ PA558913
Description
Immunogen sequence: TPKPRENNVE SQKHSLTSQS QISPKSWGVA TASLIPNDQL LPRKLNTEPK DVPKPVHQPV GSSSTLPKDP VLRKEKLQDL MTQIQGT Highest antigen sequence identity to the following orthologs: Mouse - 75%, Rat - 74%.
The protein encoded by this gene may regulate the transport of mRNA. It may play a role in the differentiation of erythroblasts. Multiple transcript variants encoding different isoforms have been found for this gene.
Specifications
CAPRIN2 | |
Polyclonal | |
Unconjugated | |
Caprin2 | |
C1q domain containing 1; C1q domain-containing protein 1; C1QDC1; caprin family member 2; caprin2; caprin-2; Caprin-2-like protein; Cytoplasmic activation/proliferation-associated protein 2; EEG1; EEG-1; gastric cancer multidrug resistance-associated protein; KIAA1873; Protein EEG-1; RNA granule protein 140; RNG140 | |
Rabbit | |
Antigen Affinity Chromatography | |
RUO | |
65981 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Liquid |
Immunohistochemistry (Paraffin), Immunocytochemistry | |
0.05 mg/mL | |
PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 | |
Q6IMN6 | |
Caprin2 | |
Recombinant protein corresponding to Human CAPRIN2. Recombinant protein control fragment (Product #RP-97680). | |
100 μL | |
Primary | |
Human | |
Antibody | |
IgG |
Safety and Handling
WARNING: Cancer - www.P65Warnings.ca.gov
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction