Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ CAPRIN2 Polyclonal Antibody
Rabbit Polyclonal Antibody
Supplier: Invitrogen™ PA570779
Description
This target displays homology in the following species: Dog: 93%; Horse: 92%; Human: 100%; Pig: 79%.
The protein encoded by this gene may regulate the transport of mRNA. It may play a role in the differentiation of erythroblasts. Multiple transcript variants encoding different isoforms have been found for this gene.
Specifications
CAPRIN2 | |
Polyclonal | |
Unconjugated | |
Caprin2 | |
C1q domain containing 1; C1q domain-containing protein 1; C1QDC1; caprin family member 2; caprin2; caprin-2; Caprin-2-like protein; Cytoplasmic activation/proliferation-associated protein 2; EEG1; EEG-1; gastric cancer multidrug resistance-associated protein; KIAA1873; Protein EEG-1; RNA granule protein 140; RNG140 | |
Rabbit | |
Affinity chromatography | |
RUO | |
65981 | |
-20°C, Avoid Freeze/Thaw Cycles | |
Liquid |
Western Blot | |
0.5 mg/mL | |
PBS with 2% sucrose and 0.09% sodium azide | |
Q6IMN6 | |
Caprin2 | |
synthetic peptide directed towards the N-terminal region of human CAPRIN2. | |
100 μL | |
Primary | |
Human | |
Antibody | |
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction