Learn More
Invitrogen™ CAPRIN2 Polyclonal Antibody

Description
Immunogen sequence: QVNHSQHGES QRALSPLQST LSSAASPSQA YETYIENGLI CLKHKIRNIE KKKLKLEDYK DRLKSGEHLN PDQLEAVEKY EEVLHNLEFA KELQKTFSGL SLDLLKAQKK AQRREHMLKL EAEKKKLRTI LQVQYVLQNL TQEHVQKDFK GGLNGAVYLP SKELDYLIKF SKLTCPERNE SLSVEDQMEQ SSLYFWDLLE GSEKAVVGTT YKHLKDLLSK LLNSGYFESI PVPKNAKEKE VP.
Specifications
Specifications
| Antigen | CAPRIN2 |
| Applications | ELISA, Western Blot |
| Classification | Polyclonal |
| Concentration | 0.96 mg/mL |
| Conjugate | Unconjugated |
| Formulation | PBS with 50% glycerol and 0.01% thimerosal; pH 7.3 |
| Gene | Caprin2 |
| Gene Accession No. | Q6IMN6 |
| Gene Alias | C1q domain containing 1; C1q domain-containing protein 1; C1QDC1; caprin family member 2; caprin2; caprin-2; Caprin-2-like protein; Cytoplasmic activation/proliferation-associated protein 2; EEG1; EEG-1; gastric cancer multidrug resistance-associated protein; KIAA1873; Protein EEG-1; RNA granule protein 140; RNG140 |
| Gene Symbols | Caprin2 |
| Show More |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.