Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

CAPRIN2 Rabbit anti-Human, Polyclonal, Bioss
SDP

Catalog No. 501982230
Encompass_Preferred
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
50-198-2230 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 50-198-2230 Supplier Bioss Supplier No. BS12423R
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

The protein encoded by this gene may regulate the transport of mRNA. It may play a role in the differentiation of erythroblasts. Multiple transcript variants encoding different isoforms have been found for this gene.

Specifications

Antigen CAPRIN2
Applications Flow Cytometry, Immunofluorescence, Immunohistochemistry (Paraffin), Western Blot
Classification Polyclonal
Concentration 1 μg/mL
Conjugate Unconjugated
Formulation PBS with 1% BSA, 50% glycerol and 0.09% sodium azide; pH
Gene Caprin2
Gene Accession No. Q6IMN6
Gene Alias C1q and tumor necrosis factor related protein 3-like; C1q domain containing 1; C1q domain-containing protein 1; C1QDC1; caprin family member 2; Caprin2; Caprin-2; Cytoplasmic activation/proliferation-associated protein 2; EEG1; EEG-1; Gastric cancer multidrug resistance-associated protein; Kiaa1873; LOC100351407; Protein EEG-1; RNA granule protein 140; Rng140
Gene Symbols Caprin2
Host Species Rabbit
Immunogen KLH conjugated synthetic peptide derived from human CAPRIN2.
Purification Method Protein A
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 65981
Target Species Human
Content And Storage -20°C
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.