Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

epsilon-Caprolactam, 99+%

Catalog No. p-3735908
Click to view available options
Quantity:
100 g
250 g
1 kg
2.5 kg
Packaging:
Plastic bottle

CAS: 105-60-2 | C6H11NO | 113.16 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 105-60-2
Molecular Formula C6H11NO
Molecular Weight (g/mol) 113.16
MDL Number MFCD00006936
InChI Key JBKVHLHDHHXQEQ-UHFFFAOYSA-N
Synonym epsilon-caprolactam, caprolactam, 6-caprolactam, 2-oxohexamethylenimine, aminocaproic lactam, 2-azacycloheptanone, hexahydro-2h-azepin-2-one, 2h-azepin-2-one, hexahydro, 6-hexanelactam, hexanolactam
PubChem CID 7768
ChEBI CHEBI:28579
IUPAC Name azepan-2-one
SMILES C1CCC(=O)NCC1

Specifications

Melting Point 68°C to 71°C
Color White
Boiling Point 268°C
Flash Point 152°C
Infrared Spectrum Authentic
Assay Percent Range 99% min. (GC)
Packaging Plastic bottle
Quantity 100 g
Beilstein 21, V,6, 444
Fieser 09,316
Merck Index 15, 1763
Solubility Information Solubility in water: 4560g/L (20°C). Other solubilities: freely soluble in methanol,dimethylformamide,,ethanol,ether and tetrahydrofurfuryl alcohol,soluble in chlorinated hydrocarbons,cyclohexene,petroleum fractions,chloroform,benzene
Viscosity 8.52 mPa.s (80°C)
Formula Weight 113.16
Percent Purity 99+%
Physical Form Crystals or Flakes
Chemical Name or Material ε-Caprolactam
Show More Show Less
Hazard Category GHS Signal Word: Warning
Hazard Statement GHS H Statement:
Causes serious eye irritation.
Causes skin irritation.
May cause respiratory irritation.
Harmful if swallowed.
Harmful if inhaled.
Precautionary Statement GHS P Statement:
Wear protective gloves/protective clothing/eye protection/face protection.
IF INHALED: Remove victim to fresh air and keep at rest in a position comfortable for breathing.
IF ON SKIN: Wash with plenty of soap and water.
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.
EINECSNumber 203-313-2

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.