Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ CADPS2 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA566013
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA566013 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA566013 Supplier Invitrogen™ Supplier No. PA566013
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Immunogen sequence: ALPKVPSLPLN LPQIPNISTA SWMPSLYEST NGSA Highest antigen sequence identity to the following orthologs - mouse 83%, rat 86%.

CAPS2 and the related protein CAPS1 encode novel neural/endocrine-specific cytosolic and peripheral membrane proteins. Both are essential components of the synaptic vesicle priming machinery and are required for the Ca2+-regulated exocytosis of secretory vesicles; CAPS-deficient neurons contain no or very few fusion competent synaptic vesicles, causing a selective impairment of fast phasic transmitter release. Like CAPS1, numerous isoforms of CAPS2 are known to exist. These isoforms show distinct expression patterns in the brain. For example, CAPS2b showed high expression in the developing cerebellum. Furthermore, one version of CAPS2 mRNA that lacks exon 3 is detected in some autistic patients, suggesting that the differential expression pattern of CAPS2 is involved in neuronal development.
TRUSTED_SUSTAINABILITY

Specifications

Antigen CADPS2
Applications Immunocytochemistry
Classification Polyclonal
Concentration 0.05 mg/mL
Conjugate Unconjugated
Formulation PBS with 40% glycerol and 0.02% sodium azide; pH 7.2
Gene CADPS2
Gene Accession No. Q86UW7
Gene Alias A230044C21Rik; Ca++-dependent secretion activator 2; Ca2+ dependent secretion activator 2; Ca2+-dependent activator protein for secretion 2; Cadps2; calcium dependent secretion activator 2; calcium-dependent activator protein for secretion 2; Calcium-dependent secretion activator 2; calcyphosin-2; calcyphosine 2; Calcyphosine-2; calcyphosphine 2; Caps2; CAPS-2; cerebellum postnatal development associated protein 2; Cpd2; KIAA1591; RGD1559440; UG0636c06
Gene Symbols CADPS2
Host Species Rabbit
Immunogen Recombinant Human CAPS2. Recombinant protein control fragment (Product #RP-106697).
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 93664
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.