Learn More
Invitrogen™ CADPS2 Polyclonal Antibody

Description
Immunogen sequence: MLDPSSSEEE SDEGLEEESR DVLVAAGSSQ RAPPAPTREG RRDAPGRAGG GGAARSVSPS PSVLSEGRDE PQRQLDDEQE.
Specifications
Specifications
| Antigen | CADPS2 |
| Applications | ELISA, Western Blot |
| Classification | Polyclonal |
| Concentration | 0.86 mg/mL |
| Conjugate | Unconjugated |
| Formulation | PBS with 50% glycerol and 0.01% thimerosal; pH 7.3 |
| Gene | CADPS2 |
| Gene Accession No. | Q86UW7, Q8BYR5 |
| Gene Alias | A230044C21Rik; Ca++-dependent secretion activator 2; Ca2+ dependent secretion activator 2; Ca2+-dependent activator protein for secretion 2; Cadps2; calcium dependent secretion activator 2; calcium-dependent activator protein for secretion 2; Calcium-dependent secretion activator 2; calcyphosin-2; calcyphosine 2; Calcyphosine-2; calcyphosphine 2; Caps2; CAPS-2; cerebellum postnatal development associated protein 2; Cpd2; KIAA1591; RGD1559440; UG0636c06 |
| Gene Symbols | CADPS2 |
| Show More |
Safety and Handling
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.