Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Capsaicin, Natural, MP Biomedicals

Catalog No. ICN202757 Shop All MP Biomedicals, Inc Products
Click to view available options
Quantity:
100 mg

Chemical Identifiers

CAS 404-86-4
Molecular Formula C18H27NO3
Molecular Weight (g/mol) 305.418
InChI Key YKPUWZUDDOIDPM-SOFGYWHQSA-N
Synonym capsaicin, zostrix, capsaicine, e-capsaicin, qutenza, styptysat, axsain, isodecenoic acid vanillylamide, trans-8-methyl-n-vanillyl-6-nonenamide
PubChem CID 1548943
ChEBI CHEBI:3374
IUPAC Name (E)-N-[(4-hydroxy-3-methoxyphenyl)methyl]-8-methylnon-6-enamide
SMILES CC(C)C=CCCCCC(=O)NCC1=CC(=C(C=C1)O)OC

Specifications

Quantity 100 mg
Formula Weight 305.4
Chemical Name or Material Capsaicin, Natural
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.