Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Capsid Protein Rabbit anti-Virus, Polyclonal, Bioss
SDP

Catalog No. 501980653
Encompass_Preferred
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
50-198-0653 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 50-198-0653 Supplier Bioss Supplier No. BS10057R
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

There has been much uncertainty in the relationship between PCV2 and PMWS but Bolin et al demonstrated that Koch′s postulates could be fulfilled by inoculating cesarean-derived, colostrum-deprived (CDCD) pigs with PCV2 and producing clinical signs of PMWS. Similar work has been performed using an infectious genomic clone of PCV2 in specific pathogen free (SPF) pigs to produce the gross and microscopic lymphoid lesions of PMWS. Although PCV2 has been shown to induce PMWS lesions in CDCD and SPF pigs, the presence of additional factors are required.

Specifications

Antigen Capsid Protein
Applications Immunofluorescence, Immunohistochemistry (Paraffin), Western Blot
Classification Polyclonal
Concentration 1 μg/mL
Conjugate Unconjugated
Formulation PBS with 1% BSA, 50% glycerol and 0.09% sodium azide; pH
Host Species Rabbit
Immunogen KLH conjugated synthetic peptide derived from PCV2 capsid protein.
Purification Method Protein A
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Target Species Virus
Content And Storage -20°C
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.