Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ CAPSL Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA561337
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA561337 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA561337 Supplier Invitrogen™ Supplier No. PA561337
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Immunogen sequence: GIKGLGRVFR IMDDDNNRTL DFKEFMKGLN DYAVVMEKEE VEELFRRFDK DG Highest antigen sequence identity to the following orthologs: Mouse - 90%, Rat - 92%.

CAPSL gene ontology annotations related to this gene include calcium ion binding.
TRUSTED_SUSTAINABILITY

Specifications

Antigen CAPSL
Applications Immunohistochemistry (Paraffin)
Classification Polyclonal
Concentration 0.4 mg/mL
Conjugate Unconjugated
Formulation PBS with 40% glycerol and 0.02% sodium azide; pH 7.2
Gene Capsl
Gene Accession No. Q8WWF8
Gene Alias 1700028N11Rik; AW125205; Calcyphosine; calcyphosine like; calcyphosine-like; calcyphosin-like protein; CAPSL; RGD1308776
Gene Symbols Capsl
Host Species Rabbit
Immunogen Recombinant protein corresponding to Human CAPSL. Recombinant protein control fragment (Product #RP-100435).
Purification Method Antigen Affinity Chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 133690
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.