Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ CAPSL Polyclonal Antibody
Rabbit Polyclonal Antibody
Supplier: Invitrogen™ PA563579
Description
Immunogen sequence: PMSRARKEVI MQAFRKLDKT GDGVITIEDL REVYNAKHHP KYQNGEWSEE Q Highest antigen sequence identity to the following orthologs: Mouse - 96%, Rat - 96%.
CAPSL gene ontology annotations related to this gene include calcium ion binding.
Specifications
CAPSL | |
Polyclonal | |
Unconjugated | |
Capsl | |
1700028N11Rik; AW125205; Calcyphosine; calcyphosine like; calcyphosine-like; calcyphosin-like protein; CAPSL; RGD1308776 | |
Rabbit | |
Antigen affinity chromatography | |
RUO | |
133690 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Liquid |
Immunohistochemistry (Paraffin), Immunocytochemistry | |
0.1 mg/mL | |
PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 | |
Q8WWF8 | |
Capsl | |
Recombinant protein corresponding to Human CAPSL. Recombinant protein control fragment (Product #RP-101822). | |
100 μL | |
Primary | |
Human | |
Antibody | |
IgG |
Safety and Handling
WARNING: Cancer - www.P65Warnings.ca.gov
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction