Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Captan 98.0+%, TCI America™
SDP

Catalog No. C205925G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
C205925G 25 g
C20595G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. C205925G Supplier TCI America Supplier No. C205925G
Add to Cart
Add to Cart

Chemical Identifiers

CAS 133-06-2
Molecular Formula C9H8Cl3NO2S
Molecular Weight (g/mol) 300.578
MDL Number MFCD00041811
InChI Key LDVVMCZRFWMZSG-UHFFFAOYSA-N
Synonym captan, amercide, orthocide, vondcaptan, captaf, captane, captex, kaptan, malipur, merpan
PubChem CID 8606
ChEBI CHEBI:34608
IUPAC Name 2-(trichloromethylsulfanyl)-3a,4,7,7a-tetrahydroisoindole-1,3-dione
SMILES C1C=CCC2C1C(=O)N(C2=O)SC(Cl)(Cl)Cl

Specifications

Melting Point 175°C
Color White
Quantity 25 g
UN Number 2811
Formula Weight 300.58
Percent Purity ≥98.0% (HPLC,N)
Physical Form Crystalline Powder
Chemical Name or Material Captan
EINECSNumber (9)-0934
RTECSNumber GW5075000
TSCA Yes
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.