Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Cytiva Capto™ Adhere Media

Catalog No. 45002962 Shop All Cytiva Products
Change view
Click to view available options
Quantity:
25 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
45-002-962 25 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 45-002-962 Supplier Cytiva Supplier No. 17544410
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

A multimodal anion exchanger to follow the Protein A capture step, making for a streamlined process

This product is designed to allow for a two-step process with purification of monoclonal antibodies and Protein A capture, which cuts cost and time.

  • Two-step chromatographic process with protein A affinity
  • Multimodal anion exchanger for post-protein A purification of monoclonal antibodies
  • Wide operational window of pH and conductivity
  • High capacity and productivity in flow-through mode
  • Average particle size of 75μm
  • Highy crossed-linked agarose
  • Ligand N-benzyl-n-methyl ethanolamine

For intermediate purification and polishing of monoclonal antibodies after the Protein A capture step.

TRUSTED_SUSTAINABILITY

Specifications

Quantity 25 mL
For Use With (Application) For Multimodal Chromatography
Stability 1 M Acetic Acid, 1 M Sodium Hydroxide
Storage Requirements 4°C to 30°C
Product Type Capto Adhere Media
Flow Rate 600 cm/Hoμr
Particle Size 75 μm
Height (English) Bed 7.87 in.
Height (Metric) Bed 20 cm
Ionic Capacity 0.09 to 0.12 mM Cl-/mL
Includes Certificate of Analysis
Ion Exchange Type Multimodal Strong AnIon Exchanger
Max. Pressure 300 kPa
Matrix Highly Cross-Linked Agarose
pH Range 2 to 14
Storage Buffer 20% Ethanol
Ligand Type N-benzyl-n-methyl ethanolamine
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.