Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Cytiva Capto™ Core 700

Catalog No. 45002595 Shop All Cytiva Products
Change view
Click to view available options
Quantity:
25 mL
100 mL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
45-002-595 25 mL
45-002-596 100 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 45-002-595 Supplier Cytiva Supplier No. 17548101
Add to Cart
Add to Cart

Capto Core 700 is designed for intermediate purification and polishing of viruses and other large biomolecules.

The novel core bead technology and multimodal, octylamine ligand give Capto Core 700 dual functionality, size exclusion, and binding chromatography in one chromatography medium (resin).

  • For intermediate purification and polishing of viruses and other large biomolecules (Mr > 700 000) in flowthrough mode.
  • Novel core bead technology allows efficient capture of contaminants while target molecules are collected in the flowthrough.
  • Significantly improved productivity and higher flow rates compared with gel filtration methods.
  • Straightforward optimization due to flowthrough chromatography and robust performance.
  • Convenient small-scale purification, process development, and scale-up using prepacked HiTrap and HiScreen columns.
  • Capto Core 700 is composed of a ligand-activated core and inactive shell.
  • The inactive shell excludes large molecules (cut off ∼ Mr 700000 [700 kDa]) from entering the core through the pores of the shell.
  • These larger molecules are collected in the column flowthrough while smaller impurities bind to the internalized ligands.
  • The core of each bead is functionalized with ligands that are both hydrophobic and positively charged, resulting in a highly efficient multimodal binding of various contaminants small enough to enter the core.
  • The multimodal ligands ensure strong binding with most impurities over a wide range of pH and salt concentrations.
  • Bound impurities are removed from the beads by cleaning-in-place (CIP) procedures using NaOH and in most cases, a solvent.

TRUSTED_SUSTAINABILITY

Specifications

Buffer 1M NaOH, 6M Guanidine Hydrochloride, 30% Isopropanol and 70% Ethanol
Product Type Capto Core 700 chromato graphy media
Quantity 25 mL
Flow Rate <500 cm/hr.
Ligand Type Octylamine
pH Range 2 to 14
Particle Size 90μm
Support Type Highly Cross-linked Agarose
Content And Storage 4° to 30°C
For Use With (Application) Intermediate purification and polishing of viruses and other large biomolecules
Matrix Multimodal Resin
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.