Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Cytiva Capto™ Core 700

Capto Core 700 is designed for intermediate purification and polishing of viruses and other large biomolecules.

$379.00 - $1015.00

Specifications

Buffer 1M NaOH, 6M Guanidine Hydrochloride, 30% Isopropanol and 70% Ethanol
Product Type Capto Core 700 chromato graphy media
Flow Rate <500cm/hr.
Ligand Type Octylamine
pH Range 2 to 14
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
45-002-596
View Documents
Cytiva
17548102
100mL
Each for $1,015.00
Add to Cart
 
45-002-595
View Documents
Cytiva
17548101
25mL
Each for $379.00
Add to Cart
 
Description

Description

The novel core bead technology and multimodal, octylamine ligand give Capto Core 700 dual functionality, size exclusion, and binding chromatography in one chromatography medium (resin).

  • For intermediate purification and polishing of viruses and other large biomolecules (Mr > 700 000) in flowthrough mode.
  • Novel core bead technology allows efficient capture of contaminants while target molecules are collected in the flowthrough.
  • Significantly improved productivity and higher flow rates compared with gel filtration methods.
  • Straightforward optimization due to flowthrough chromatography and robust performance.
  • Convenient small-scale purification, process development, and scale-up using prepacked HiTrap and HiScreen columns.
  • Capto Core 700 is composed of a ligand-activated core and inactive shell.
  • The inactive shell excludes large molecules (cut off ∼ Mr 700000 [700 kDa]) from entering the core through the pores of the shell.
  • These larger molecules are collected in the column flowthrough while smaller impurities bind to the internalized ligands.
  • The core of each bead is functionalized with ligands that are both hydrophobic and positively charged, resulting in a highly efficient multimodal binding of various contaminants small enough to enter the core.
  • The multimodal ligands ensure strong binding with most impurities over a wide range of pH and salt concentrations.
  • Bound impurities are removed from the beads by cleaning-in-place (CIP) procedures using NaOH and in most cases, a solvent.

Specifications

Specifications

1M NaOH, 6M Guanidine Hydrochloride, 30% Isopropanol and 70% Ethanol
<500cm/hr.
2 to 14
Highly Cross-linked Agarose
Intermediate purification and polishing of viruses and other large biomolecules
Capto Core 700 chromato graphy media
Octylamine
90μm
4° to 30°C
Multimodal Resin
SDS
Documents

Documents

Product Certifications
Provide Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.