Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Cytiva Capto™ MMC ImpRes Multimodal BioProcess™ Chromatography Medium

Catalog No. 45002506 Shop All Cytiva Products
Change view
Click to view available options
Quantity:
25 mL
100 mL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
45-002-506 25 mL
45-002-507 100 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 45-002-506 Supplier Cytiva Supplier No. 17371601
Add to Cart
Add to Cart

For high-resolution polishing of monoclonal antibodies (MAbs) and other biomolecules

  • High yields achieved through high-resolution beads and ligand selectivity
  • Efficient removal of aggregates, viruses, and main contaminants in MAb processes
  • Enables use of platform approach to MAb process development
  • Displays broad pH/salt operational window

Avoid using oxidizing agents, cationic detergents.

TRUSTED_SUSTAINABILITY

Specifications

Quantity 25 mL
For Use With (Application) For High Resolution Polishing of Monoclonal Antibodies (MAbs) and Other Biomolecules
Stability 1 M Acetic Acid, 1 M NaOH, 2 M NaCl, 5% 1-Propanol, 30% Isopropanol, 70% Ethanol
Storage Requirements 4°C to 30°C
Anion Exchange Capacity 25 to 39 μM/mL
Binding Capacity 60 to 90 mg Mab/mL
Product Type Capto MMC ImpRes Media
Particle Size 40 μm
Max. Pressure 300 kPa
Matrix High-flow Agarose
pH Range 3 to 12
Storage Buffer 20% Ethanol, 0.2 M Sodium Acetate
Ligand Type Multimodal Weak CatIon Exchanger
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.