Learn More
Globe Scientific™ CapTrack™ Recapping Archiver with Position Database
Globe Scientific™ CapTrack™ Recapping Archiver with Position Database dramatically increases the efficiency, safety and accuracy of blood lab processing procedures.
Manufacturer: Globe Scientific™ CT2
Description
- Portable benchtop device that automates blood tubes recapping, tracking and sample management for a small to mid size blood processing labs
Specifications
50/60 Hz | |
14.2 in. | |
90 lb. | |
20.7 in. |
22.5 x 20.7 x 14.2 in. | |
22.5 in. | |
120/240 V |