Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Thermo Scientific™ CaptureSelect™ Factor X Affinity Matrix

Catalog No. p-7180766
Click to view available options
Quantity:
5 mL
10 mL
50 mL
250 mL
1 L

Thermo Scientific™ CaptureSelect™ Factor X Affinity Matrix was specifically designed for the purification of human Factor X from plasma and recombinant sources.

CaptureSelect Factor X Affinity Matrix was specifically designed for the purification of human Factor X from plasma and recombinant sources.

Features of the CaptureSelect Factor X Affinity Matrix include:

  • Highly selective for human Factor X with no affinity for other human plasma proteins
  • Binds both Factor X and Factor Xa in the presence of calcium
  • Enables efficient purification of Factor X with high yields and retained activity
  • Mild elution at neutral pH using EDTA
  • Offers excellent scalability
  • Non-animal-derived

Coagulation Factor X is a vitamin K-dependent, liver-produced serine protease that serves as the first enzyme in the coagulation cascade to form fibrin. It is a two-chain glycoprotein with a molecular weight of approximately 59 kDa. Factor X normally circulates in the plasma as an inactive molecule with activation of Factor X involved in both the intrinsic and extrinsic coagulation pathways.

Free of animal components

CaptureSelect products contain affinity ligands based on recombinant single-domain antibody fragments (VHH) created by a proprietary technology. The VHH affinity ligand is a 12 to 15 kDa fragment comprising the three complementarity-determining regions (CDRs) that form the antigen-binding domain, efficiently produced in the yeast Saccharomyces cerevisiae by a production process free of any animal components (Animal Origin-Free).

Main characteristics

  • Matrix: agarose-based
  • Average particle size: 65 ± 10 μm
  • Ligand: CaptureSelect Factor X affinity ligand
  • Ligand coupling method: epoxide
  • Binding capacity: >10 mg/mL resin
  • Binding conditions: 50 mM Tris, 150 mM NaCl, 5 mM CaCl2, pH 7
  • Elution conditions: 50 mM Tris, 150 mM NaCl, 5 mM EDTA, pH 7
  • Flow characteristics: 50–200 cm/h (up to 2 bar)
  • Formulation buffer: 20% (v/v) ethanol

Specifications

Binding Capacity >10 mg/mL
Flow Rate 50 to 200 cm/hr. (Up to 2 bar)
Matrix Agarose-based
Content And Storage Store in refrigerator (2°C to 8°C).
Shipping Condition Wet Ice
Particle Size 65 ±10 μm
Form Agarose Bead
Product Line CaptureSelect™
Quantity 5 mL
Target Protein
Type Affinity Matrix
Ligand Type Factor X affinity ligand
Stationary Phase Factor X
Column Type Agarose Resin
Show More Show Less
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.