Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Thermo Scientific™ CaptureSelect™ FcXP Ligand Leakage ELISA
GREENER_CHOICE

Catalog No. 810371201
Change view
Click to view available options
Quantity:
1 assay x 96-well plate
10 assay x 96-well plate
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
81-037-1201 1 assay x 96-well plate
81-037-1210 10 assay x 96-well plate
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 81-037-1201 Supplier Thermo Scientific™ Supplier No. 810371201
Only null left
Add to Cart
Add to Cart

Thermo Scientific™ CaptureSelect™ FcXP Ligand Leakage ELISA is designed to detect possible leached ligand of antibodies or antibody fragments purified using CaptureSelect FcXP Affinity Matrix.

CaptureSelect FcXP Ligand Leakage ELISA is designed to detect possible leached ligand of antibodies or antibody fragments purified using CaptureSelect FcXP Affinity Matrix. The FcXP affinity ligand is a 13-kD recombinant protein, expressed in yeast and covalently coupled to a chromatography matrix for high chemical stability and low leakage. During the chromatography process small amounts of the affinity ligand can leach off of the chromatography support and co-elute with the protein.

This ELISA provides typical sensitivity and accurate quantitation of possible leached ligand greater than or equal to 1 ng/mL. The CaptureSelect FcXP Ligand Leakage ELISA can be used as a tool to aid in optimal purification process development and in routine quality control of in-process streams as well as final product.

Specifications

Content And Storage 3 x 0.1 mL, store at -20°C.
Shipping Condition Dry Ice
Form Reagent Solution
Product Line CaptureSelect
Quantity 1 assay x 96-well plate
Target CaptureSelect FcXP Ligand
Type ELISA Kit
Sufficient For 1 x 96-well plate

For Research Use Only. Not for use in diagnostic procedures.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.