Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Thermo Scientific™ CaptureSelect™ FcXP MiniChrom™ Pre-packed Column
GREENER_CHOICE

Catalog No. 5943712005
Change view
Click to view available options
Volume (Metric):
1 mL
5 mL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Volume (Metric)
59-437-12005 5 mL
59-437-12001 1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 59-437-12005 Supplier Thermo Scientific™ Supplier No. 5943712005
Only null left
Add to Cart
Add to Cart

Thermo Scientific™ CaptureSelect™ FcXP MiniChrom™ columns are designed for bench-scale resin screening, process development, and sample preparation.

CaptureSelect FcXP MiniChrom™ columns are designed for bench-scale resin screening, process development, and sample preparation.

These pre-packed columns are ready to connect and have the following features:

  • Dimensions: 0.8 x 2 cm (1 mL), 0.8 x 10 cm (5 mL)
  • Direct connection to standard chromatography systems
  • Packed with CaptureSelect FcXP Affinity Matrix, offering high dynamic binding capacity

The 1-mL columns can be used for quick screenings of application feasibility and lab scale purification on a convenient and easy-to-use pre-packed column. The 10-cm bed height of the 5-mL columns allows initial process development on a bench scale.

CaptureSelect FcXP Affinity Matrix recognizes the CH3 domain of all human IgG subclasses (IgG1, 2, 3, and 4), enabling purification of all IgG subclasses and Fc fusion proteins directly from complex source materials in a robust and efficient purification process with excellent purity obtained in a single step.

Features of the FcXP affinity matrix include:

  • Mild elution, making it suitable for Fc fusion proteins
  • One-step purification from crude material with high purity and yield
  • Human specific, no binding to bovine antibodies
  • Excellent scalability
  • Non–animal-derived

Free of animal components

CaptureSelect products contain affinity ligands based on recombinant single-domain antibody fragments (VHH) created by a proprietary technology. The VHH affinity ligand is a 12 to 15 kDa fragment comprising the three complementarity-determining regions (CDRs) that form the antigen-binding domain, efficiently produced in the yeast Saccharomyces cerevisiae by a production process free of any animal components (Animal Origin-Free).

Main characteristics

  • Matrix: agarose-based, epoxide-activated
  • Average particle size: 65 ± 10 μm
  • Ligand: CaptureSelect FcXP affinity ligand
  • Ligand coupling: Epoxide coupling
  • Binding capacity: 40-50 g IgG/L resin depending on flow rate, column height, and contact time
  • Elution conditions: 20 mM citric acid or acetic acid, pH 3–4
  • Column dimensions: 0.8 x 2 cm (1 mL), 0.8 x 10 cm (5 mL)
  • Flow characteristics: 75–150 cm/h (up to 2 bar)
  • Formulation buffer: 20% (v/v) ethanol

Specifications

Product Line CaptureSelect
Type Pre-packed Column
Quantity 5 mL
Column Type MiniChrom
Particle Size 65 ±10 μm
Binding Capacity 40 to 50 g IgG/L resin
Flow Rate 75 to 150 cm/hr. (Up to 2 bar)
Ligand Type FcXP affinity ligand
Matrix Agarose-based, epoxide-activated
Volume (Metric) 5 mL
Diameter (Metric) 0.8 cm
Length (Metric) 10 cm
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.