Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Thermo Scientific™ CaptureSelect™ IgG-Fc (rabbit) Affinity Matrix

Catalog No. p-7180785
Click to view available options
Quantity:
5 mL
10 mL
50 mL

Thermo Scientific™ CaptureSelect™ IgG-Fc (Rabbit) Affinity Matrix has been specifically designed for the purification of rabbit IgG from recombinant and plasma sources.

CaptureSelect IgG-Fc (Rabbit) Affinity Matrix has been specifically designed for the purification of rabbit IgG from recombinant and plasma sources. The ligand on the resin targets the Fc region of rabbit IgG antibodies independent of the light chain subclass.

Features of this affinity matrix include:

  • Highly selective for rabbit IgG
  • No binding to mouse, rat, human, bovine, horse, sheep, and goat IgG
  • Complex growth media with fetal calf or horse serum can be applied
  • Non-animal-derived

The high selectivity and yields obtained using CaptureSelect IgG-Fc (Rabbit) Affinity Matrix enables a robust and efficient purification process with excellent purity obtained in one step.

Free of animal components

CaptureSelect products contain affinity ligands based on recombinant single-domain antibody fragments (VHH) created by a proprietary technology. The VHH affinity ligand is a 12 to 15 kDa fragment comprising the three complementarity-determining regions (CDRs) that form the antigen-binding domain, efficiently produced in the yeast Saccharomyces cerevisiae by a production process free of any animal components (Animal Origin-Free).

Main characteristics:

  • Matrix: agarose-based
  • Average particle size: 65 ± 10 μm
  • Ligand: CaptureSelect IgG-Fc (rabbit) affinity ligand
  • Ligand coupling method: epoxide
  • Binding capacity: >15 mg rabbit IgG/mL resin
  • Elution conditions: 1 M glycine, pH 3.0, or 20 mM acetic or citric acid, pH 3.0
  • Flow characteristics: 50–200 cm/h (up to 2 bar)
  • Formulation buffer: 20%(v/v) ethanol

Specifications

Binding Capacity >15 mg rabbit IgG/mL resin
Flow Rate 50 to 200 cm/hr. (Up to 2 bar)
Matrix Agarose-based
Content And Storage Store in refrigerator (2°C to 8°C).
Shipping Condition Wet Ice
Particle Size 65 ±10 μm
Form Liquid Suspension
Product Line CaptureSelect™
Quantity 5 mL
Type Affinity Matrix
Ligand Type IgG-Fc (rabbit) affinity ligand
Stationary Phase Anti-IgG-Fc (rabbit)
Show More Show Less
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.