Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
                
Learn More
Learn More
                                    CAPZA1 Antibody (OTI2G4) - Azide and BSA Free, Novus Biologicals™
                                    
                                    
                                    
                                    
                                
                            
                            
                            
                            
Mouse Monoclonal Antibody
Supplier: Novus Biologicals NBP270336
Description
CAPZA1 Monoclonal specifically detects CAPZA1 in Human, Mouse, Rat samples. It is validated for Western Blot, Flow Cytometry, Immunohistochemistry, CyTOF-ready.Specifications
| CAPZA1 | |
| Monoclonal | |
| LYOPH | |
| Western Blot 1:500, Flow Cytometry 1:100, Immunohistochemistry 1:50, CyTOF-ready | |
| Cap Z, CAPPA1, capping protein (actin filament) muscle Z-line, alpha 1, CAPZ, CapZ alpha-1, CAZ1, F-actin-capping protein subunit alpha-1 | |
| Mouse | |
| 32.7 kDa | |
| 100 μg | |
| Primary | |
| Reconstitute with PBS (pH 7.3). To use this carrier-free antibody for conjugation experiments, another round of desalting is highly recommended. | |
| Store at -20°C. Avoid freeze-thaw cycles. | |
| IgG1 | 
| Western Blot, Flow Cytometry, Immunohistochemistry, CyTOF | |
| OTI2G4 | |
| Unconjugated | |
| Lyophilized from PBS (pH 7.3) with 8% Trehalose with No Preservative | |
| CAPZA1 | |
| Full length human recombinant protein of human CAPZA1(NP_006126) produced in HEK293T cell. | |
| Affinity Purified | |
| Extracellular Matrix | |
| 829 | |
| Human, Mouse, Rat | |
| Lyophilized | 
                    Product Content Correction
                
                Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
			
            Spot an opportunity for improvement?Share a Content Correction